Anti-LPAR4 Rabbit Polyclonal Antibody (Cy7®)
Lieferant: Bioss
BS-12074R-CY7
BOSSBS-12074R-CY7EA 900 EUR
BOSSBS-12074R-CY7
Anti-LPAR4 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Receptor for lysophosphatidic acid (LPA), a mediator of diverse cellular activities. Transduces a signal by increasing the intracellular calcium ions and by stimulating adenylyl cyclase activity. The rank order of potency for agonists of this receptor is 1-oleoyl- >1-stearoyl- >1-palmitoyl- >1-myristoyl- >1-alkyl- >1-alkenyl-LPA (By similarity).
Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200
Type: Primary
Antigen: LPAR4
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat