Anti-GABARE Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss
BS-12081R-CY5
BOSSBS-12081R-CY5EA 900 EUR
BOSSBS-12081R-CY5
Anti-GABARE Rabbit Polyclonal Antibody (Cy5®)
Antikörper
GAD-65 and GAD-67, glutamate decarboxylases, function to catalyze the production of GABA (gamma-aminobutyric acid). In the central nervous system GABA functions as the main inhibitory transmitter by increasing a Cl- conductance that inhibits neuronal firing. GABA has been shown to activate both ionotropic (GABAA) and metabotropic (GABAB) receptors as well as a third class of receptors called GABAC. Both GABAA and GABAC are ligand-gated ion channels, however, they are structurally and functionally distinct. Members of the GABAA receptor family include GABAA R Alpha 1-6, GABAA R Beta 1-3, GABAA R Gamma 1-3, GABAA R Delta, GABAA R Epsilon, GABAA R Zeta 1 and GABAA R Zeta 2. The GABAB family is composed of GABAB R1 Alpha and GABAB R1 Beta. GABA transporters have also been identified and include GABA T-1, GABA T-2 and GABA T-3 (also designated GAT-1, -2, and -3). The GABA transporters function to terminate GABA action.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GABARE/GABA Receptor Epsilon
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR