Anti-NMDAR3A + 3B Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss
BS-12103R-A647
BOSSBS-12103R-A647EA 1350 EUR
BOSSBS-12103R-A647
Anti-NMDAR3A + 3B Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Glutamate receptors mediate most excitatory neurotransmission in the brain and play an important role in neural plasticity, neural development and neuro-degeneration. Ionotropic glutamate receptors are categorized into NMDA receptors and kainate/AMPA receptors, both of which contain glutamate-gated, cation-specific ion channels. Kainate/AMPA receptors co-localize with NMDA receptors in many synapses and consist of seven structurally related subunits designated GluR-1 to 7. The kainate/AMPA receptors are primarily responsible for fast excitatory neurotransmission by glutamate, whereas the NMDA receptors exhibit slow kinesis of Ca2+ ions and a high permeability for Ca2+ ions. One such NMDA receptor, NR3B, is expressed in motor neurons and forms cation channels impermeable to calcium, which can resist many open-channel blockers. NR3B functions in the brain as an excitatory glycine receptor, modifying the normal role of glycine as an inhibitory neurotransmitter.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NMDAR3A + 3B
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR