Anti-ACCN4 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-12129R-A555
BOSSBS-12129R-A555EA 893 EUR
BOSSBS-12129R-A555
Anti-ACCN4 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
This gene belongs to the superfamily of acid-sensing ion channels, which are proton-gated, amiloride-sensitive sodium channels. These channels have been implicated in synaptic transmission, pain perception as well as mechanoperception. This gene is predominantly expressed in the pituitary gland, and was considered a candidate for paroxysmal dystonic choreoathetosis (PDC), a movement disorder, however, no correlation was found between mutations in this gene and PDC. [provided by RefSeq, Feb 2012].

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: ACCN4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR