Anti-ZIC5 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-12147R-CY7
BOSSBS-12147R-CY7EA 900 EUR
BOSSBS-12147R-CY7
Anti-ZIC5 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Zic5 (zinc finger protein of the cerebellum 5) is a C2H2 zinc finger transcription factor that influences development of the neural crest. Zic family members are abundant in developing and adult cerebellum. Zic family members are important during development, and have been associated with X-linked visceral heterotaxy and holoprosencephaly type 5. Zic5 is closely linked to Zic2, a related family member on chromosome 13.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ZIC5
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR