Anti-ZIC5 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Lieferant: Bioss
BS-12147R-HRP
BOSSBS-12147R-HRPEA 900 EUR
BOSSBS-12147R-HRP
Anti-ZIC5 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Zic5 (zinc finger protein of the cerebellum 5) is a C2H2 zinc finger transcription factor that influences development of the neural crest. Zic family members are abundant in developing and adult cerebellum. Zic family members are important during development, and have been associated with X-linked visceral heterotaxy and holoprosencephaly type 5. Zic5 is closely linked to Zic2, a related family member on chromosome 13.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500
Type: Primary
Antigen: ZIC5
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat