Anti-CYB5R3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss
BS-12162R-HRP
BOSSBS-12162R-HRPEA 900 EUR
BOSSBS-12162R-HRP
Anti-CYB5R3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
CYB5R3 is a 301 amino acid protein encoded by the human gene CYB5R3. CYB5R3 belongs to the flavoprotein pyridine nucleotide cytochrome reductase family and has two naturally occuring isoforms. Isoform 1 is anchored to the cytoplasmic side of the endoplasmic reticulum membrane and mitochondrion outer membrane, while isoform 2 is the soluble form found in erythrocytes. CYB5R3 is involved in the desaturation and elongation of fatty acids, cholesterol biosynthesis, drug metabolism and, in erythrocytes, methemoglobin reduction. A serine residue at position 117 seems to only be found in persons of African origin. The allele frequency is 0.23 in African Americans. It is not found in Caucasians, Asians, Indo-Aryans or Arabs. This difference seems to have no effect on the enzyme activity. Defects in CYB5R3 are the cause of hereditary methemoglobinemia (HM). There are three forms of this disease: type 1 (HM1), in which the enzyme is only deficient in erythrocytes with a mild cyanosis; type 2 (HM2), in which the enzyme is completely deficient; and type 3 (HM3), where the deficiency is seen in all blood cells. Type 2 is a severe form accompanied by mental retardation and neurological impairment.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: CYB5R3
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR