Anti-CAPS2 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-12163R-CY3
BOSSBS-12163R-CY3EA 900 EUR
BOSSBS-12163R-CY3
Anti-CAPS2 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
CAPS2 is a 557 amino acid calcium-binding protein that is abundantly expressed, with highest expression found in placenta, testis, colon, lung and brain. CAPS2 contains three EF-hand domains and exists as three alternatively spliced isoforms. Suggested to play a role in large dense-core vesicle (LDCV) exocytosis, CAPS2 is encoded by a gene that maps to human chromosome 12, which encodes over 1,100 genes and comprises approximately 4.5% of the human genome. Chromosome 12 is associated with a variety of diseases and afflictions, including hypochondrogenesis, achondrogenesis, Kniest dysplasia, Noonan syndrome and trisomy 12p, which causes facial developmental defects and seizure disorders.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CAPS2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR