Anti-KCNT2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-12177R-A680
BOSSBS-12177R-A680EA 890 EUR
BOSSBS-12177R-A680
Anti-KCNT2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Voltage-gated K+ channels in the plasma membrane are important regulators of electrical Signalling, controlling the repolarisation and the frequency of action potentials in neurons, muscles and other excitable cells. KCNT2 is a 1,135 amino acid multi-pass transmembrane protein belonging to the potassium channel family (calcium-activated subfamily) of proteins. KCNT2 produces rapidly activating outward rectifier potassium currents in reponse to high intracellular sodium and chloride levels. Its channel activity is inhibited by ATP, inhalation anesthetics, such as isoflourane, and upon stimulation of G-protein coupled receptors, such as mAChR M1 and GluR-1. There are four isoforms of KCNT2 that are produced as a result of alternative splicing events.

Type: Primary
Antigen: KCNT2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 51-150/1135
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR