Anti-KCNT2 Rabbit Polyclonal Antibody (Cy3®)
Lieferant: Bioss
BS-12177R-CY3
BOSSBS-12177R-CY3EA 900 EUR
BOSSBS-12177R-CY3
Anti-KCNT2 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Voltage-gated K+ channels in the plasma membrane are important regulators of electrical signaling, controlling the repolarization and the frequency of action potentials in neurons, muscles and other excitable cells. KCNT2 is a 1,135 amino acid multi-pass transmembrane protein belonging to the potassium channel family (calcium-activated subfamily) of proteins. KCNT2 produces rapidly activating outward rectifier potassium currents in reponse to high intracellular sodium and chloride levels. Its channel activity is inhibited by ATP, inhalation anesthetics, such as isoflourane, and uponMore Product Information