Anti-HOXB6 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-12194R
BOSSBS-12194REA 725 EUR
BOSSBS-12194R
Anti-HOXB6 Rabbit Polyclonal Antibody
Antikörper
HoxB6 is cytoplasmically expressed throughout fetal epidermal development, but displays nuclear expression in normal adult skin. HoxB6 protein also has nuclear expression in hyperproliferative skin conditions, but appears to be localized in the cytoplasm in basal and squamous cell carcinomas. HoxB6 genes are also expressed in normal adult lung.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: HOXB6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR