Anti-ZNF346 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-12235R-A750
BOSSBS-12235R-A750EA 890 EUR
BOSSBS-12235R-A750
Anti-ZNF346 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. ZNF346 (Zinc finger protein 346), also known as JAZ (Just another zinc finger protein), is a 294 amino acid protein that contains four matrin-type zinc fingers. The matrin-type zinc finger, which is very similar in structure to the classical DNA-binding C2H2 zinc finger, was first identified in the protein matrin-3. The matrin-type zinc finger has also been identified in several spliceosome RNA-binding proteins. Two isoforms exists due to alternate splicing events.

Type: Primary
Antigen: ZNF346
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 181-294/294
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR