Anti-ZNF449 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-12236R-A680
BOSSBS-12236R-A680EA 890 EUR
BOSSBS-12236R-A680
Anti-ZNF449 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a Kr_ppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. As a member of the krueppel C2H2-type zinc-finger protein family, ZNF449 is a 518 amino acid protein that contains one SCAN box domain and seven C2H2-type zinc fingers. ZNF449 is ubiquitously expressed and localizes to the nucleus. There are three isoforms of ZNF449 that are produced as a result of alternative splicing events.

Type: Primary
Antigen: ZNF449
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 401-518/518
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR