Anti-ZNF449 Rabbit Polyclonal Antibody (Cy5®)
Lieferant: Bioss
BS-12236R-CY5
BOSSBS-12236R-CY5EA 900 EUR
BOSSBS-12236R-CY5
Anti-ZNF449 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a Krüppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. As a member of the krueppel C2H2-type zinc-finger protein family, ZNF449 is a 518 amino acid protein that contains one SCAN box domain and seven C2H2-type zinc fingers. ZNF449 is ubiquitously expressed and localizes to the nucleus. There are three isoforms of ZNF449 that are produced as a result of alternative splicing events.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ZNF449
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat