Anti-ZNF704 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-12241R-CY3
BOSSBS-12241R-CY3EA 900 EUR
BOSSBS-12241R-CY3
Anti-ZNF704 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a Krüppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. As a member of the krueppel C2H2-type zinc-finger protein family, ZNF704 is a 412 amino acid nuclear protein that contains one C2H2-type zinc finger. The gene encoding ZNF704 maps to human chromosome 8, which is made up of nearly 146 million bases and encodes about 800 genes. Chromosome 8 is also associated with Pfeiffer syndrome, congenital hypothyroidism and Waardenburg syndrome.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ZNF704
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR