Anti-SNX7 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss
BS-12250R-FITC
BOSSBS-12250R-FITCEA 900 EUR
BOSSBS-12250R-FITC
Anti-SNX7 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Sorting nexin (SNX) proteins are members of a large family of hydrophilic PX (phospholipid-binding motif) domain-containing proteins that interact with a variety of receptor types. SNXs are widely expressed, although the tissue distribution of each SNX mRNA varies. The ability of SNXs to bind specific phospholipids, as well as their tendency to form protein-protein complexes, suggests a role for these proteins in cellular membrane trafficking and protein sorting. SNXs may also function specifically in pro-degradative sorting, internalization, endosomal recycling or simply in endosomal sorting. SNXs partially associate with cellular membranes, despite their hydrophilic nature. SNX7 is unique in that it does not have a coiled coil region like some of the SNX family members. Mutations in the SNX7 gene have not been shown to cause any diseases.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SNX7
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR