Anti-Fox2/RBM9 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-12299R-A680
BOSSBS-12299R-A680EA 890 EUR
BOSSBS-12299R-A680
Anti-Fox2/RBM9 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
RNA-binding protein that regulates alternative splicing events by binding to 5'-UGCAUGU-3' elements. Prevents binding of U2AF2 to the 3'-splice site. Regulates alternative splicing of tissue-specific exons and of differentially spliced exons during erythropoiesis (By similarity). RNA-binding protein that seems to act as a coregulatory factor of ER-alpha.

Type: Primary
Antigen: Fox2/RBM9
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 65-170/390
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR