Anti-RBFOX2 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-12299R-CY3
BOSSBS-12299R-CY3EA 900 EUR
BOSSBS-12299R-CY3
Anti-RBFOX2 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
RNA-binding protein that regulates alternative splicing events by binding to 5'-UGCAUGU-3' elements. Prevents binding of U2AF2 to the 3'-splice site. Regulates alternative splicing of tissue-specific exons and of differentially spliced exons during erythropoiesis (By similarity). RNA-binding protein that seems to act as a coregulatory factor of ER-alpha.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RBFOX2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR