Anti-ENPEP Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss
BS-12307R-A488
BOSSBS-12307R-A488EA 893 EUR
BOSSBS-12307R-A488
Anti-ENPEP Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Appears to have a role in the catabolic pathway of the renin-angiotensin system. Probably plays a role in regulating growth and differentiation of early B-lineage cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ENPEP
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR