Anti-TRIM22 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-12330R
BOSSBS-12330REA 725 EUR
BOSSBS-12330R
Anti-TRIM22 Rabbit Polyclonal Antibody
Antikörper
The tripartite motif (TRIM) family of proteins are characterized by a conserved TRIM domain that includes a coiled-coil region, a B-box type zinc finger, one RING finger and three zinc-binding domains. Staf-50 (50 kDa-stimulated trans-acting factor), also known as TRIM22 (tripartite motif-containing 22), RNF94 or GPSTAF50, is a 498 amino acid cytoplasmic protein that belongs to the TRIM family and, characteristic of TRIM family members, contains one RING-type zinc finger, one B box-type zinc finger and one SPRY domain. Induced by IFN-å and IFN-∫, Staf-50 is strongly expressed in ovary, spleen, thymus and peripheral blood leukocytes where it is thought to mediate the antiviral effects of IFN proteins. Additionally, Staf-50 is present in leukemic cells, suggesting a role in cancer formation and metastasis. Staf-50 exists as two alternatively spliced isoforms which are encoded by a gene that maps to human chromosome 11.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: TRIM22
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR