Anti-HFE/Hemochromatosis Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-12335R-A680
BOSSBS-12335R-A680EA 874 EUR
BOSSBS-12335R-A680
Anti-HFE/Hemochromatosis Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
The features of hemochromatosis include cirrhosis of the liver, diabetes, hypermelanotic pigmentation of the skin, and heart failure. Since hemochromatosis is a relatively easily treated disorder if diagnosed, this is a form of preventable cancer. The HFE protein, which is defective in hereditary hemo-chromatosis, normally is expressed in crypt enterocytes of the duodenum where it has a unique, predominantly intracellular localisation. In placenta, the HFE protein co-localizes with and forms a stable association with the transferrin receptor (TfR), providing a link between the HFE protein and iron transport. Immunocytochemistry shows that the HFE protein and TfR both are expressed in the crypt enterocytes. Western blots show that, as is the case in human placenta, the HFE protein in crypt enterocytes is physically associated with the TfR and with 2-microglobulin. It is proposed that HFE has two mutually exclusive activities in cells: inhibition of uptake or inhibition of release of iron and that the balance between serum transferrin saturation and serum transferrin-receptor concentrations determines which of these functions predominates. The gene which encodes HFE maps to human chromosome 6p21.3.

Type: Primary
Antigen: HFE/Hemochromatosis
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 262-348/348
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR