Anti-IPMK Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Lieferant: Bioss
BS-12385R-A555
BOSSBS-12385R-A555EA 893 EUR
BOSSBS-12385R-A555
Anti-IPMK Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Inositol polyphosphate multikinase (IPMK) belongs to the inositol phosphokinase (IPK) family and is characterized as having a broad substrate specificity. However, IPMK displays a preference for inositol-1,4,5-trisphosphate (Ins(1,4,5)P3) and inositol 1,3,4,6-tetrakisphosphate (Ins(1,3,4,6)P4). IPMK is ubiquitously expressed with the highest expression in skeletal muscle, liver, placenta, lung, peripheral blood leukocytes, kidney, spleen and colon. IPMK is localized to the nucleus, where it may play a role in the regulation of calcium release from intracellular stores and has been implicatedMore Product Information