Anti-SNX4 Rabbit Polyclonal Antibody (Cy7®)
Lieferant: Bioss
BS-12406R-CY7
BOSSBS-12406R-CY7EA 900 EUR
BOSSBS-12406R-CY7
Anti-SNX4 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
SNX2 and SNX4 share a high degree of amino acid similarity with SNX1, as they all contain a characteristic phox homology (PX) domain (4). These proteins are all partially associated with cellular membranes, and they, likewise, associate with EGF, PDGF and insulin receptor tyrosine kinases (2). These nexins are widely expressed and yet have various tissue distribution patterns. Additionally, the sorting nexins can associate with each other and with a variety of other cellular proteins, suggesting that they exist as part of multisubunit complexes (1,5). The related protein, SNX3, comprises a distinct subgroup of nexins that share less sequence similarity outside of the PX domain and have dramatically different binding affinities for the tyrosine kinase receptors (2,6).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SNX4
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat