Anti-SNX9 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-12407R
BOSSBS-12407REA 725 EUR
BOSSBS-12407R
Anti-SNX9 Rabbit Polyclonal Antibody
Antikörper
Sorting nexin (SNX) proteins are members of a large family of hydrophilic PX (phospholipid-binding motif) domain-containing proteins that interact with a variety of receptor types. SNXs are widely expressed, although the tissue distribution of each SNX mRNA varies. The ability of SNXs to bind specific phospholipids, as well as their tendency to form protein-protein complexes, suggests a role for these proteins in cellular membrane trafficking and protein sorting. SNXs may also function specifically in pro-degradative sorting, internalization, endosomal recycling or simply in endosomal sorting. SNXs partially associate with cellular membranes, despite their hydrophilic nature. SNX9 resides in the cytosol where it influences the processing and trafficking of insulin receptors. The enzyme aldolase binds to and inactivates SNX9. Phosphorylation of SNX9 releases aldolase and frees SNX9 to recruit and activate Dynamin II, a neuronal phosphoprotein and a GTPase enzyme which mediates late stages of endocytosis in both neural and non-neural cells.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: SNX9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR