Anti-BAMBI Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-12418R-A555
BOSSBS-12418R-A555EA 901 EUR
BOSSBS-12418R-A555
Anti-BAMBI Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
BAMBI is a membrane spanning glycoprotein that acts as a negative regulator of TGF-beta signaling during development. The BAMBI family of proteins are related to type I TGF-beta receptor family, however, BAMBI is a pseudoreceptor that lacks an intracellular serine/threonine kinase domain. BAMBI transcription regulation is under the influence of beta-catenin, BMP, smad3 and smad4. BAMBI expression can increase in colorectal and hepatocellular carcinomas relative to non-cancerous tissues. BAMBI is expressed at high levels during odontogenesis. BAMBI is coexpressed with Bmp-4 during early Xenopus embryogenesis and can be detected in poorly metastatic human melanoma cell lines.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: BAMBI
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR