Anti-DOCK2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss
BS-12431R-A488
BOSSBS-12431R-A488EA 893 EUR
BOSSBS-12431R-A488
Anti-DOCK2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Involved in cytoskeletal rearrangements required for lymphocyte migration in response of chemokines. Activates RAC1 and RAC2, but not CDC42, by functioning as a guanine nucleotide exchange factor (GEF), which exchanges bound GDP for free GTP. May also participate in IL2 transcriptional activation via the activation of RAC2.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DOCK2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR