Anti-LRRFIP1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss
BS-12439R-A647
BOSSBS-12439R-A647EA 893 EUR
BOSSBS-12439R-A647
Anti-LRRFIP1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
LRRFIP1 is an 738 amino acid human protein whose rodent counterpart is known as Lrrfip1 (also designated FLAP in mouse). LRRFIP1 is also believed to control smooth muscle cell proliferation following arterial injury through PDGF-A repression. The N-terminus of LRRFIP1 shows high homology to the coiled-coil domain of FLAP, a protein which binds the leucine-rich repeat (LRR) of Flightless I, and the interaction of LRRFIP1 with the LRR of Flightless I has been confirmed. LRRFIP1 does not bind single-stranded DNA or RNA significantly and binds double-stranded DNA weakly. In contrast, LRRFIP1 binds double-stranded RNA with high affinity, and two molecules of LRRFIP1 bind the TaR stem. The RNA binding domain has been identified and encompasses a lysine-rich motif. Flightless I has a C-terminal TaR-like domain which binds Actin and therefore the association of LRRFIP1 with the LRR of Flightless I may provide a link between the Actin cytoskeleton and RNA in mammalian cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LRRFIP1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR