Anti-ABCF2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
BS-12441R-A750
BOSSBS-12441R-A750EA 890 EUR
BOSSBS-12441R-A750
Anti-ABCF2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
ATP-binding cassette (ABC) transporters are an evolutionarily conserved family of widely-expressed proteins that use ATP hydrolysis to catalyse the transport of various molecules across extracellular and intracellular membranes. As the largest family of transmembrane proteins, ABC genes comprise several subfamilies. Eukaryotic ABC transporters are largely responsible for trafficking hydrophobic compounds either within the cell, as part of a metabolic process, or outside the cell, for transport to other organs or for secretion from the body. ABCF2 in particular plays a putative role in tumour suppression at metastatic sites and in the endocrine pathway for breast cancer and may be a prognostic marker for clear cell ovarian adenocarcinoma.
Type: Primary
Antigen: ABCF2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 301-400/623
Host: Rabbit
Isotype: IgG
Reactivity: Rat