Anti-ABCF2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss
BS-12441R-HRP
BOSSBS-12441R-HRPEA 900 EUR
BOSSBS-12441R-HRP
Anti-ABCF2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
ATP-binding cassette (ABC) transporters are an evolutionarily conserved family of widely-expressed proteins that use ATP hydrolysis to catalyze the transport of various molecules across extracellular and intracellular membranes. As the largest family of transmembrane proteins, ABC genes comprise several subfamilies. Eukaryotic ABC transporters are largely responsible for trafficking hydrophobic compounds either within the cell, as part of a metabolic process, or outside the cell, for transport to other organs or for secretion from the body. ABCF2 in particular plays a putative role in tumor suppression at metastatic sites and in the endocrine pathway for breast cancer and may be a prognostic marker for clear cell ovarian adenocarcinoma.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: ABCF2
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR