Anti-ADH5 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-12448R-A555
BOSSBS-12448R-A555EA 893 EUR
BOSSBS-12448R-A555
Anti-ADH5 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
The alcohol dehydrogenase family of proteins metabolize a wide variety of substrates, including retinol, hydroxysteroids, ethanol, aliphatic alcohols and lipid peroxidation products. ADH5 (alcohol dehydrogenase 5 (class III)), also known as FDH (formaldehyde dehydrogenase), ADHX, ADH-3 or GSNOR, is a 374 amino acid cytoplasmic protein that belongs to the class III subfamily of alcohol dehydrogenases. Expressed ubiquitously, ADH5 uses iron as a cofactor to catalytically oxidize both long-chain primary alcohols and S-hydroxymethyl-glutathione, a product formed spontaneously between formaldehyde and glutathione. ADH5 exists as a homodimer and, via its ability to oxidize S-hydroxymethyl-glutathione and, thus, eliminate formaldehyde, functions as an important component of cellular metabolism. Genetic variations in the gene encoding ADH5 may affect drug and alcohol dependence in humans.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ADH5
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR