Anti-ADH6 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Lieferant: Bioss
BS-12449R-A555
BOSSBS-12449R-A555EA 893 EUR
BOSSBS-12449R-A555
Anti-ADH6 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
ADH6 (alcohol dehydrogenase 6), also known as ADH-5, is a 368 amino acid member of the class V zinc-containing alcohol dehydrogenase family. This family of enzymes functions to metabolize a wide variety of substrates such as retinol, hydroxysteroids, ethanol, aliphatic alcohols and lipid peroxidation products. Localized to the cytoplasm and expressed in the stomach and liver, ADH6 catalyzes the reversible oxidation of alcohols to their corresponding aldehydes or ketones and is able to bind two zinc ions as cofactors. ADH6 contains a glucocorticoid response element upstream of its 5' UTRMore Product Information