Anti-AIPL1 Rabbit Polyclonal Antibody (ALEXA FLUOR® 750)

Lieferant: Bioss
BS-12452R-A750
BOSSBS-12452R-A750EA 890 EUR
BOSSBS-12452R-A750
Anti-AIPL1 Rabbit Polyclonal Antibody (ALEXA FLUOR® 750)
Antikörper
The inherited blindness associated protein, aryl hydrocarbon receptor interacting protein-like 1 (AIPL1), interacts with the cell cycle regulator protein NUB1. AIPL1 is crucial for protein folding and stabilisation, as well as for protein trafficking. It localises to the nucleus or cytoplasm and is highly expressed in the pineal gland and the retina. In the retina, AIPL1 is expressed in both developing cone and rod photoreceptors, but it is restricted to rod photoreceptors in the adult human retina. Defects in the gene encoding for AIPL1 can cause Leber congenital amaurosis type IV, an early-onset, inherited autosomal recessive disorder that results in childhood blindness.

Type: Primary
Antigen: AIPL1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 165-260/384
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR