Anti-AIPL1 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-12452R-CY3
BOSSBS-12452R-CY3EA 900 EUR
BOSSBS-12452R-CY3
Anti-AIPL1 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
The inherited blindness associated protein, aryl hydrocarbon receptor interacting protein-like 1 (AIPL1), interacts with the cell cycle regulator protein NUB1. AIPL1 is crucial for protein folding and stabilization, as well as for protein trafficking. It localizes to the nucleus or cytoplasm and is highly expressed in the pineal gland and the retina. In the retina, AIPL1 is expressed in both developing cone and rod photoreceptors, but it is restricted to rod photoreceptors in the adult human retina. Defects in the gene encoding for AIPL1 can cause Leber congenital amaurosis type IV, an early-onset, inherited autosomal recessive disorder that results in childhood blindness.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: AIPL1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR