Anti-ALDH9A1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-12461R-A680
BOSSBS-12461R-A680EA 890 EUR
BOSSBS-12461R-A680
Anti-ALDH9A1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Aldehyde dehydrogenases (ALDHs) mediate the NADP+-dependent oxidation of aldehydes into acids and play an important role in the detoxification of alcohol-derived acetaldehyde, as well as in lipid peroxidation and in the metabolism of corticosteroids, biogenic amines and neurotransmitters. ALDH9A1 (aldehyde dehydrogenase family 9 member A1), also known as E3, ALDH4, ALDH7, ALDH9 or TMABADH, is a 494 amino acid cytoplasmic protein that is highly expressed in adult liver, skeletal muscle, kidney and embryonic brain. ALDH9A1 converts gamma-trimethylaminobutyraldehyde into gamma-butyrobetaine and catalyses the irreversible oxidation of a broad range of aldehydes to the corresponding acids in a NAD-dependent reaction.

Type: Primary
Antigen: ALDH9A1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 401-494/494
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR