Anti-APC Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss
BS-12481R-A647
BOSSBS-12481R-A647EA 921 EUR
BOSSBS-12481R-A647
Anti-APC Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Tumor suppressor. Promotes rapid degradation of CTNNB1 and participates in Wnt signaling as a negative regulator. APC activity is correlated with its phosphorylation state. Activates the GEF activity of SPATA13 and ARHGEF4. Plays a role in hepatocyte growth factor (HGF)-induced cell migration. Required for MMP9 up-regulation via the JNK signaling pathway in colorectal tumor cells. Acts as a mediator of ERBB2-dependent stabilization of microtubules at the cell cortex. It is required for the localization of MACF1 to the cell membrane and this localization of MACF1 is critical for its function in microtubule stabilization.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: APC
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR