Anti-OBP/Aphrodisin Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-12488R-A680
BOSSBS-12488R-A680EA 890 EUR
BOSSBS-12488R-A680
Anti-OBP/Aphrodisin Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
This protein is found in nasal epithelium and it binds a wide variety of chemical odorants.

Type: Primary
Antigen: OBP/Aphrodisin
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 51-150/172
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR