Anti-OBP/Aphrodisin Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
BS-12488R-A750
BOSSBS-12488R-A750EA 890 EUR
BOSSBS-12488R-A750
Anti-OBP/Aphrodisin Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
This protein is found in nasal epithelium and it binds a wide variety of chemical odorants.
Type: Primary
Antigen: OBP/Aphrodisin
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 51-150/172
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen