Anti-OBP1F Rabbit Polyclonal Antibody (Cy7®)
Lieferant: Bioss
BS-12488R-CY7
BOSSBS-12488R-CY7EA 900 EUR
BOSSBS-12488R-CY7
Anti-OBP1F Rabbit Polyclonal Antibody (Cy7®)
Antikörper
This protein is found in nasal epithelium and it binds a wide variety of chemical odorants.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: OBP1F
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat