Anti-ARF1 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-12508R-CY3
BOSSBS-12508R-CY3EA 900 EUR
BOSSBS-12508R-CY3
Anti-ARF1 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
GTP-binding protein that functions as an allosteric activator of the cholera toxin catalytic subunit, an ADP-ribosyltransferase. Involved in protein trafficking among different compartments. Modulates vesicle budding and uncoating within the Golgi complex. Deactivation induces the redistribution of the entire Golgi complex to the endoplasmic reticulum, suggesting a crucial role in protein trafficking. In its GTP-bound form, its triggers the association with coat proteins with the Golgi membrane. The hydrolysis of ARF1-bound GTP, which is mediated by ARFGAPs proteins, is required for dissociation of coat proteins from Golgi membranes and vesicles.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ARF1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR