Anti-ACTRT3 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss
BS-12529R-A647
BOSSBS-12529R-A647EA 893 EUR
BOSSBS-12529R-A647
Anti-ACTRT3 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Actin-related proteins are classified into Arp subclasses according to their amino acid sequence similarity to Actin. Both Arps and Actin proteins have an ATPase domain, which catalyzes the decomposition of adenosine triphosphate (ATP) into adenosine diphosphate (ADP) and a free phosphate ion to release energy. ArpM1 (actin-related protein M1) is a 372 amino acid ubiquitously expressed protein that localizes to both the cytoplasm and the cytoskeleton and belongs to the Arp family of Actin-like proteins. The gene encoding ArpM1 maps to human chromosome 3, which houses over 1,100 genes, including a chemokine receptor (CKR) gene cluster and a variety of human cancer-related gene loci.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ACTRT3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR