Anti-ATF6B Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-12543R-CY7
BOSSBS-12543R-CY7EA 900 EUR
BOSSBS-12543R-CY7
Anti-ATF6B Rabbit Polyclonal Antibody (Cy7®)
Antikörper
ATF-6 is a member of the basic-leucine zipper family of transcription factors. Endoplasmic reticulum stress causes cleavage of transmembrane ATF-6 and transclocation of active ATF-6 to the nucleus. Soluble ATF-6 can exist as either an ATF-6beta homodimer or an ATF-6Alpha/beta heterodimer. Binding of the ATF-6beta homodimer or ATF-6Alpha/beta heterodimer to the nuclear transcription factor Y C (NF-YC) induces ER chaperone transcription.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ATF6B
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR