Anti-FGF3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss
BS-1255R-A350
BOSSBS-1255R-A350EA 876 EUR
BOSSBS-1255R-A350
Anti-FGF3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Plays an important role in the regulation of embryonic development, cell proliferation, and cell differentiation. Required for normal ear development (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FGF3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR