Anti-HTRA2 Rabbit Polyclonal Antibody (Cy5®)
Lieferant: Bioss
BS-1271R-CY5
BOSSBS-1271R-CY5EA 900 EUR
BOSSBS-1271R-CY5
Anti-HTRA2 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Serine protease that shows proteolytic activity against a non-specific substrate beta-casein. Promotes or induces cell death either by direct binding to and inhibition of BIRC proteins(also called inhibitor of apoptosis proteins, IAPs), leading to an increase in caspase activity, or by a BIRC inhibition-independent, caspase-independent and serine protease activity-dependent mechanism. Isoform 2 seems to be proteolytically inactive. [CATALYTIC ACTIVITY] Cleavage of non-polar aliphatic amino-acids at the P1 position, with a preference for Val, Ile and Met. At the P2 and P3 positions, Arg isMore Product Information