Anti-HTRA2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss
BS-1271R-HRP
BOSSBS-1271R-HRPEA 900 EUR
BOSSBS-1271R-HRP
Anti-HTRA2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Serine protease that shows proteolytic activity against a non-specific substrate beta-casein. Promotes or induces cell death either by direct binding to and inhibition of BIRC proteins(also called inhibitor of apoptosis proteins, IAPs), leading to an increase in caspase activity, or by a BIRC inhibition-independent, caspase-independent and serine protease activity-dependent mechanism. Isoform 2 seems to be proteolytically inactive. [CATALYTIC ACTIVITY] Cleavage of non-polar aliphatic amino-acids at the P1 position, with a preference for Val, Ile and Met. At the P2 and P3 positions, Arg is selected most strongly with a secondary preference for other hydrophilic residues. [SUBUNIT] Homotrimer. Interacts with MXI2. The mature protein, but not the precursor, binds to BIRC2, BIRC3 and XIAP.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: HTRA2
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR