Anti-8-OHdG/8 Hydroxyguanosine Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-1278R
BOSSBS-1278REA 698 EUR
BOSSBS-1278R
Anti-8-OHdG/8 Hydroxyguanosine Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: 8-OHdG/8 Hydroxyguanosine
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Others
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR