Anti-BTC Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss
BS-12864R-A350
BOSSBS-12864R-A350EA 893 EUR
BOSSBS-12864R-A350
Anti-BTC Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Betacellulin (BTC), a member of the epidermal growth factor (EGF) family, was originally identified as a growth-promoting factor in the conditioned medium of a mouse pancreatic-cell carcinoma (insulinoma) cell line and has since been identified in humans. BTC is synthesized as a large transmembrane precursor molecule that can be cleaved proteolytically to release the soluble form of BTC or function as membrane-anchored growth factors in juxtacrine signaling. BTC, in addition to stimulating homodimers of ErbB-1 and ErbB-4, is capable of binding and activating all possible combinations of heterodimeric ErbB receptors including the oncogenic ErbB-2/ErbB-3 complex. BTC is also expressed in some human malignancies and may have an important role in tumor growth progression.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: BTC
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR