Anti-ARFGEF1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss
BS-12866R-FITC
BOSSBS-12866R-FITCEA 900 EUR
BOSSBS-12866R-FITC
Anti-ARFGEF1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Guanine nucleotide-exchange proteins (GEPs) accelerate replacement of bound GDP with GTP and thereby activate ADP-ribosylation factors (ARFs), a family of guanine nucleotide-binding proteins that play an important role in intracellular vesicular trafficking. GEPs comprise two major families, large GEPs that are inhibited by brefeldin A (BFA), a protein that effects Golgi structure and a group of smaller GEPs that are insenstive to BFA. Two genes for GEPs found on human chromosomes 8 and 20 encode BFA sensitive GEPs designated BIG1 and BIG2. Both GEPS contain a sec7 domain that is responsible for their brefeldin inhibition and also their catalytic activity. In vivo, BIG1 and BIG2 exist in macromolecular complexes that move between the Golgi membranes and cytosol. BIG2 associates with PKA regulatory subunits, implying that BIG2 may act as an A kinase-anchoring protein (AKAP) that could coordinate the cAMP and ARF regulatory pathways.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ARFGEF1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR