Anti-BLVRA Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss
BS-12868R-A488
BOSSBS-12868R-A488EA 893 EUR
BOSSBS-12868R-A488
Anti-BLVRA Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
In human liver cytosolic fractions, four forms of biliverdin reductase have been identified, including two biliverdin-IX Beta reductases and two biliverdin-IX Alpha reductases, designated isozymes I and II and isozymes III and IV, respectively. Biliverdin reductase A (BLVRA), also designated biliverdin-IX Alpha-reductase, belongs to the GFO/iIDH/MocA family and the biliverdin reductase subfamily. The gene that encodes this cytoplasmic protein maps to chromosome 7p14-cen. BLVRA reduces biliverdin IX ?(the ?methene bridge of the open tetrapyrrole) to bilirubin with the concomitant oxidation of an NADH or NADPH cofactor (bilirubin + NADP+ = biliverdin + NADPH). BLVRA is expressed primarily in liver.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BLVRA
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR