Anti-BLVRA/Biliverdin Reductase Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
BS-12868R-A750
BOSSBS-12868R-A750EA 890 EUR
BOSSBS-12868R-A750
Anti-BLVRA/Biliverdin Reductase Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
In human liver cytosolic fractions, four forms of biliverdin reductase have been identified, including two biliverdin-IX Beta reductases and two biliverdin-IX Alpha reductases, designated isozymes I and II and isozymes III and IV, respectively. Biliverdin reductase A (BLVRA), also designated biliverdin-IX Alpha-reductase, belongs to the GFO/iIDH/MocA family and the biliverdin reductase subfamily. The gene that encodes this cytoplasmic protein maps to chromosome 7p14-cen. BLVRA reduces biliverdin IX (the methene bridge of the open tetrapyrrole) to bilirubin with the concomitant oxidation of an NADH or NADPH cofactor (bilirubin + NADP+ = biliverdin + NADPH). BLVRA is expressed primarily in liver.
Type: Primary
Antigen: BLVRA/Biliverdin Reductase
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 161-260/296
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat