Anti-NFKBIA/IKB alpha Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-1287R-A750
BOSSBS-1287R-A750EA 899 EUR
BOSSBS-1287R-A750
Anti-NFKBIA/IKB alpha Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Inhibits the activity of dimeric NF-kappa-B/REL complexes by trapping REL dimers in the cytoplasm through masking of their nuclear localisation signals. On cellular stimulation by immune and proinflammatory responses, becomes phosphorylated promoting ubiquitination and degradation, enabling the dimeric RELA to translocate to the nucleus and activate transcription.

Type: Primary
Antigen: NFKBIA/IKB alpha
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1-120/314
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine, Chicken
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR